Synonyms:
Acetic acid isopropyl ester
A0036-500ML
A0036-25ML
TCA0036-500MLEA
32.97
USD
InStock
TCA0036-500ML
TCA0036-25ML
Isopropyl acetate ≥99.0%
Isopropyl acetate
CAS Number: 108-21-4
MDL Number: MFCD00008877
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 7
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Acetic Acid Isopropyl Ester