Synonyms:
Acetic acid butyl ester
, Butyl ethanoate
, Acetic acid n-butyl ester
, n-Butyl acetate
A0228-500ML
TCA0228-500MLEA
62.93
USD
InStock
TCA0228-500ML
Butyl acetate ≥99.0%
Butyl acetate
[for Spectrophotometry]
CAS Number: 123-86-4
MDL Number: MFCD00009445
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Melting point (°C): -78
Flash Point (°C): 28
Specific Gravity (20/20): 0.88
Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Acetic Acid Butyl Ester