Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyramine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 2-(4-Hydroxyphenyl)ethylammonium chloride4-(2-Aminoethyl)phenol hydrochloride4-Hydroxyphenethylammonium chloride

A0303-25G A0303-1G
TCA0303-001GEA 20.3 USD InStock
TCA0303-001G TCA0303-025G
Tyramine hydrochloride ≥98.0% (by titrimetric analysis)
Tyramine hydrochloride
CAS Number: 60-19-5
MDL Number: MFCD00012901
Molecular Formula: C8H11NO
Molecular Weight: 173.64
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 269

Other Product Names / Synonyms: 4-Hydroxyphenethylamine Hydrochloride, 2-(4-Hydroxyphenyl)ethylamine Hydrochloride


Formula: C₈H₁₂ClNO
MW: 173.64 g/mol
Melting Pt: 269 °C
MDL Number: MFCD00012901
CAS Number: 60-19-5
EINECS: 200-462-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/30/28927730.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty