Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9,10-Anthraquinone ≥98.0%

Supplier: TCI America
Synonyms: Anthraquinone

A0502-500G A0502-25G
TCA0502-025GEA 29.41 USD InStock
TCA0502-025G TCA0502-500G
9,10-Anthraquinone ≥98.0%
9,10-Anthraquinone
CAS Number: 84-65-1
MDL Number: MFCD00001188
Molecular Formula: C14H8O2
Molecular Weight: 208.22
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 380
Melting point (°C): 284
Flash Point (°C): 185


Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 380 °C
Melting Pt: 284 °C
Density: 1.419…1.438 g/cm³ (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00001188
CAS Number: 84-65-1
EINECS: 201-549-0
Merck Index: 13,00692
https://us-legacy.vwr.com/stibo/web/std.lang.all/79/34/28927934.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty