Synonyms:
Thiazol-2-ylamine
A0633-25G
A0633-100G
A0633-500G
TCA0633-500GEA
188.73
USD
InStock
TCA0633-500G
TCA0633-025G
TCA0633-100G
2-Aminothiazole ≥98.0% (by titrimetric analysis)
2-Aminothiazole
CAS Number: 96-50-4
MDL Number: MFCD00005325
Molecular Formula: C3H4N2S
Molecular Weight: 100.14
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 90