Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A0938-25ML
TCA0938-025MLEA 269.99 USD InStock
TCA0938-025ML
4-Pentylaniline ≥97.0% (by GC, titration analysis)
4-Pentylaniline
CAS Number: 33228-44-3
MDL Number: MFCD00007926
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 265
Flash Point (°C): 113
Specific Gravity (20/20): 0.93

Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Pentylaniline


Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 265 °C
Density: 0.93 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00007926
CAS Number: 33228-44-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/86/28928686.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty