Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-5-chloro-2-methoxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: 4-Amino-5-chloro-o-anisic acid

A1237-500G A1237-25G
TCA1237-500GEA 787.59 USD InStock
TCA1237-500G TCA1237-025G
4-Amino-5-chloro-2-methoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-5-chloro-2-methoxybenzoic acid
CAS Number: 7206-70-4
MDL Number: MFCD00038794
Molecular Formula: C8H8ClNO3
Molecular Weight: 201.61
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Other Product Names / Synonyms: 4-Amino-5-chloro-2-methoxybenzoic Acid


Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 213…215 °C
MDL Number: MFCD00038794
CAS Number: 7206-70-4
EINECS: 230-582-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/91/00/28929100.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty