B0062-ZONE
TCB0062-ZONEEA
93.38
USD
InStock
TCB0062-ZONE
Benzoic acid ≥99.0%, zone refined: number of passes 20
Benzoic acid
CAS Number: 65-85-0
MDL Number: MFCD00002398
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 249
Melting point (°C): 125
Flash Point (°C): 118