Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-chloropropane ≥99.0%

Supplier: TCI America

B0575-25G B0575-500G
TCB0575-25GEA 20.12 USD InStock
TCB0575-25G TCB0575-500G
1-Bromo-3-chloropropane ≥99.0%
1-Bromo-3-chloropropane
CAS Number: 109-70-6
MDL Number: MFCD00000998
Molecular Formula: C3H6BrCl
Molecular Weight: 157.44
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Melting point (°C): -59
Specific Gravity (20/20): 1.60

Other Product Names / Synonyms: 3-Bromopropyl Chloride, 3-Chloropropyl Bromide, Trimethylene Bromochloride, Trimethylene Chlorobromide


Formula: BrH₂CCH₂CH₂Cl
MW: 157.44 g/mol
Boiling Pt: 143 °C
Melting Pt: –59 °C
Density: 1.60 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000998
CAS Number: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/16/28932516.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty