Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromoheptane ≥98.0%

Supplier: TCI America
Synonyms: 1-Heptyl bromide

B0598-500G B0598-25G
TCB0598-500GEA 90.29 USD InStock
TCB0598-500G TCB0598-25G
1-Bromoheptane ≥98.0%
1-Bromoheptane
CAS Number: 629-04-9
MDL Number: MFCD00000273
Molecular Formula: C7H15Br
Molecular Weight: 179.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Melting point (°C): -58
Flash Point (°C): 60
Specific Gravity (20/20): 1.14

Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Heptyl Bromide


Formula: BrH₂C(CH₂)₅CH₃
MW: 179.1 g/mol
Boiling Pt: 180 °C
Melting Pt: –58 °C
Density: 1.14 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000273
CAS Number: 629-04-9
EINECS: 211-068-8
UN: 1993
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/37/28932537.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty