Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-methylpropane ≥97.0%

Supplier: TCI America
Danger
Synonyms: Isobutyl bromide

B0616-500G B0616-25G
TCB0616-500GEA 76 USD InStock
TCB0616-500G TCB0616-25G
1-Bromo-2-methylpropane ≥97.0%
1-Bromo-2-methylpropane
CAS Number: 78-77-3
MDL Number: MFCD00000217
Molecular Formula: C4H9Br
Molecular Weight: 137.02
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 18
Freezing point (°C): -117
Specific Gravity (20/20): 1.27

Other Product Names / Synonyms: Isobutyl Bromide


Formula: C₄H₉Br
MW: 137.02 g/mol
Boiling Pt: 92 °C
Melting Pt: –118 °C
Density: 1.27 (20 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000217
CAS Number: 78-77-3
EINECS: 201-141-2
UN: 2342
ADR: 3,II
Merck Index: 13,05152
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/47/28932547.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty