B0734-25ML
TCB0734-025MLEA
17.86
USD
InStock
TCB0734-025ML
2-tert-Butylphenol ≥98.0%
2-tert-Butylphenol
CAS Number: 88-18-6
MDL Number: MFCD00002223
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Flash Point (°C): 102
Specific Gravity (20/20): 0.98