Synonyms:
Tribromomethane
Stabilization:
Stabilised with Stabilised
B0806-100ML
TCB0806-100MLEA
516.04
USD
InStock
TCB0806-100ML
Bromoform ≥99.0% stabilized
Bromoform
(stabilized with 2-Methyl-2-butene)
[for Spectrophotometry]
CAS Number: 75-25-2
MDL Number: MFCD00000128
Molecular Formula: CHBr3
Molecular Weight: 252.73
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Freezing point (°C): 8
Specific Gravity (20/20): 2.89
Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Tribromomethane