Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzonitrile ≥98.0%

Supplier: TCI America
Synonyms: m-Bromobenzonitrile

B1227-5G B1227-25G B1227-100G
TCB1227-100GEA 235.48 USD InStock
TCB1227-100G TCB1227-025G TCB1227-005G
3-Bromobenzonitrile ≥98.0%
3-Bromobenzonitrile
CAS Number: 6952-59-6
MDL Number: MFCD00001796
Molecular Formula: C7H4BrN
Molecular Weight: 182.02
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 40
Flash Point (°C): 110


Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 225 °C
Melting Pt: 40 °C
Density: 1.562 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001796
CAS Number: 6952-59-6
EINECS: 230-127-9
UN: 3439
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/30/60/28933060.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty