Synonyms:
Ethyl 5-bromovalerate
, 5-Bromovaleric acid ethyl ester
B1557-25G
TCB1557-025GEA
121.75
USD
InStock
TCB1557-025G
Ethyl-5-bromovalerate ≥97.0%
Ethyl 5-bromovalerate
CAS Number: 14660-52-7
MDL Number: MFCD00000266
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 227
Flash Point (°C): 104
Specific Gravity (20/20): 1.31
Other Product Names / Synonyms: 5-Bromovaleric Acid Ethyl Ester