Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fullerene C70 ≥98.0% (by HPLC)

Supplier: TCI America
Synonyms: Buckminster-Fullerene C70

B1694-100MG
TCB1694-100MGEA 172.55 USD InStock
TCB1694-100MG
Fullerene C70 ≥98.0% (by HPLC)
Fullerene C70
CAS Number: 115383-22-7
MDL Number: MFCD00146976
Molecular Formula: C70
Molecular Weight: 840.77
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal

Product Categories: Materials Chemistry, Organic Transistor Materials, Molecular Conductors, Solar Cell Research Reagents, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: Buckminsterfullerene C70, C70, Carbon C70


Formula: C₇₀
MW: 840.77 g/mol
Melting Pt: > 280 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00282904
CAS Number: 115383-22-7
UN: 1325
ADR: 4.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/34/50/28933450.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty