Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol green in ethanol (contains NaOH) stain, for TLC

Supplier: TCI America
Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

B2401-500ML
TCB2401-500MLEA 144.13 USD InStock
TCB2401-500ML
Bromocresol green in ethanol (contains NaOH) stain, for TLC
Bromocresol green
[for TLC Stain]
CAS Number: 76-60-8
MDL Number: MFCD00005874
Molecular Formula: C21H14Br4O5S
Molecular Weight: 698.01
Form: Liquid
Color: Deep Yellow Green
Flash Point (°C): 13


Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Flash Pt: 13 °C
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 263-657-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/41/40/28934140.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty