Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2-nitrobenzene ≥99.0%

Supplier: TCI America
Danger
Synonyms: 1-Chloro-2-nitrobenzene2-Chloronitrobenzene

C0220-25G C0220-500G
TCC0220-25GEA 15.63 USD InStock
TCC0220-25G TCC0220-500G
1-Chloro-2-nitrobenzene ≥99.0%
1-Chloro-2-nitrobenzene
CAS Number: 88-73-3
MDL Number: MFCD00007061
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 246
Flash Point (°C): 124
Freezing point (°C): 32

Other Product Names / Synonyms: 1-Chloro-2-nitrobenzene


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 246 °C
Melting Pt: 33 °C
Density: 1.368 g/cm³ (20 °C)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007061
CAS Number: 88-73-3
EINECS: 201-854-9
UN: 1578
ADR: 6.1,II
Merck Index: 13,02170
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/74/28937274.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty