C0231-500G
C0231-25G
TCC0231-500GEA
163.42
USD
InStock
TCC0231-500G
TCC0231-25G
4-Chloro-2-nitrotoluene
4-Chloro-2-nitrotoluene
CAS Number: 89-59-8
MDL Number: MFCD00007215
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 115
Melting point (°C): 38
Flash Point (°C): 120