Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Curcumin 0.1% in ethanol 95%

Supplier: TCI America
Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

C0435-500ML
TCC0435-500MLEA 141.09 USD InStock
TCC0435-500ML
Curcumin 0.1% in ethanol 95%
Curcumin
CAS Number: 458-37-7
MDL Number: MFCD00008365
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 13


Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Flash Pt: 13 °C
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/58/28937458.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty