C0510-100ML
C0510-25ML
C0510-500ML
TCC0510-100MLEA
33.6
USD
InStock
TCC0510-100ML
TCC0510-500ML
TCC0510-25ML
Cyclopentanone ≥99.0%
Cyclopentanone
CAS Number: 120-92-3
MDL Number: MFCD00001409
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Melting point (°C): -51
Flash Point (°C): 34
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Adipic Ketone, Ketocyclopentane, Ketopentamethylene