Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzaldehyde ≥99.0%

Supplier: TCI America
Danger

C0561-500G C0561-25G
TCC0561-025GEA 26.17 USD InStock
TCC0561-025G TCC0561-500G
2-Chlorobenzaldehyde ≥99.0%
2-Chlorobenzaldehyde
CAS Number: 89-98-5
MDL Number: MFCD00003304
Molecular Formula: C7H5ClO
Molecular Weight: 140.57
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 104
Freezing point (°C): 9
Specific Gravity (20/20): 1.25


Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 212 °C
Melting Pt: 10 °C
Density: 1.25 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00003304
CAS Number: 89-98-5
EINECS: 201-956-3
UN: 3265
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/79/28937579.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty