Synonyms:
Dimethylchlorosilane
C0778-25ML
C0778-250ML
TCC0778-025MLEA
54.79
USD
InStock
TCC0778-025ML
TCC0778-250ML
Chlorodimethylsilane ≥95.0%
Chlorodimethylsilane
CAS Number: 1066-35-9
MDL Number: MFCD00000495
Molecular Formula: C2H7ClSi
Molecular Weight: 94.61
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 37
Melting point (°C): -111
Flash Point (°C): -20
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Reduction, Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Dimethylchlorosilane, DMCS