Synonyms:
1-(Chloromethyl)-3-methylbenzene
, 3-Methylbenzyl chloride
C0865-500G
C0865-25G
TCC0865-25GEA
35.53
USD
InStock
TCC0865-25G
TCC0865-500G
α-Chloro-m-xylene ≥97.0%
α-Chloro-m-xylene
CAS Number: 620-19-9
MDL Number: MFCD00000909
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 76
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 3-Methylbenzyl Chloride