Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopropyl chloroacetate ≥98.0%

Supplier: TCI America
Danger

C0948-25ML C0948-500ML
TCC0948-25MLEA 34.48 USD InStock
TCC0948-25ML TCC0948-500ML
Isopropyl chloroacetate ≥98.0%
Isopropyl chloroacetate
CAS Number: 105-48-6
MDL Number: MFCD00040410
Molecular Formula: C5H9ClO2
Molecular Weight: 136.58
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 45
Specific Gravity (20/20): 1.09

Other Product Names / Synonyms: Chloroacetic Acid Isopropyl Ester


Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 150 °C
Melting Pt: –112 °C
Density: 1.09 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00040410
CAS Number: 105-48-6
EINECS: 203-301-7
UN: 2947
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/89/28938189.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty