Synonyms:
2-(Trifluoromethyl)benzonitrile o-(Trifluoromethyl)benzonitrile 2-Cyanobenzotrifluoride
C1004-25G
C1004-5G
TCC1004-025GEA
37.51
USD
InStock
TCC1004-025G
TCC1004-005G
α,α,α-Trifluoro-o-toluonitrile ≥98.0%
α,α,α-Trifluoro-o-toluonitrile
CAS Number: 447-60-9
MDL Number: MFCD00001791
Molecular Formula: C8H4F3N
Molecular Weight: 171.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Flash Point (°C): 90
Freezing point (°C): 19
Specific Gravity (20/20): 1.29
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Cyanobenzotrifluoride, alpha,alpha,alpha-Trifluoro-o-tolunitrile