Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodimethylvinylsilane ≥97.0%

Supplier: TCI America
Synonyms: Vinyldimethylchlorosilane Dimethylvinylchlorosilane

C1208-25ML C1208-100ML
TCC1208-025MLEA 63.95 USD InStock
TCC1208-025ML TCC1208-100ML
Chlorodimethylvinylsilane ≥97.0%
Chlorodimethylvinylsilane
CAS Number: 1719-58-0
MDL Number: MFCD00018090
Molecular Formula: C4H9ClSi
Molecular Weight: 120.65
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): -6
Specific Gravity (20/20): 0.89

Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Dimethylvinylchlorosilane, DMVS-Cl


Formula: C₄H₉ClSi
Boiling Pt: 82 °C
Density: 0.89 (20 °C)
Flash Pt: –6 °C
MDL Number: MFCD00018090
CAS Number: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/84/55/28938455.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty