Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-α-methylbenzyl alcohol ≥98.0%

Supplier: TCI America
Synonyms: 1-(3-Chlorophenyl)ethanol3-Chlorophenyl methyl carbinolm-Chloro-α-methylbenzyl alcohol

C1382-10G
TCC1382-010GEA 83.2 USD InStock
TCC1382-010G
3-Chloro-α-methylbenzyl alcohol ≥98.0%
3-Chloro-α-methylbenzyl alcohol
CAS Number: 6939-95-3
MDL Number: MFCD00021864
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 246
Flash Point (°C): 110
Specific Gravity (20/20): 1.17

Other Product Names / Synonyms: 1-(3-Chlorophenyl)ethanol, 1-(3-Chlorophenyl)ethyl Alcohol


Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 246 °C
Density: 1.17 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00021864
CAS Number: 6939-95-3
EINECS: 230-079-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/21/28938621.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty