Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-o-xylene ≥96.0%

Supplier: TCI America
Synonyms: 3-Chloro-1,2-dimethylbenzene1-Chloro-2,3-dimethylbenzene

C1558-25G
TCC1558-025GEA 72 USD InStock
TCC1558-025G
3-Chloro-o-xylene ≥96.0%
3-Chloro-o-xylene
CAS Number: 608-23-1
MDL Number: MFCD00060689
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 73
Specific Gravity (20/20): 1.08

Other Product Names / Synonyms: 3-Chloro-1,2-dimethylbenzene, 2,3-Dimethylchlorobenzene


Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 190 °C
Density: 1.08 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00060689
CAS Number: 608-23-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/78/28938778.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty