Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dibromosalicylaldehyde ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning
Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde

D0188-25G
TCD0188-025GEA 152.25 USD InStock
TCD0188-025G
3,5-Dibromosalicylaldehyde ≥98.0% (by HPLC, titration analysis)
3,5-Dibromosalicylaldehyde
CAS Number: 90-59-5
MDL Number: MFCD00003318
Molecular Formula: C7H4Br2O2
Molecular Weight: 279.92
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 83

Other Product Names / Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde


Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 83 °C
MDL Number: MFCD00003318
CAS Number: 90-59-5
EINECS: 202-003-4
Merck Index: 13,03052
https://us-legacy.vwr.com/stibo/web/std.lang.all/45/95/28974595.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty