Synonyms:
1,3-Di(dimethylethyl)-4-hydroxybenzene
D0229-25G
D0229-500G
TCD0229-500GEA
101.57
USD
InStock
TCD0229-500G
TCD0229-25G
2,4-Di-tert-butylphenol ≥97.0%
2,4-Di-tert-butylphenol
CAS Number: 96-76-4
MDL Number: MFCD00008828
Molecular Formula: C14H22O
Molecular Weight: 206.33
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 261
Melting point (°C): 56
Flash Point (°C): 128