Synonyms:
1,2-Diaminocyclohexane
, 1,2-Cyclohexanediamine
D0277-25ML
D0277-500ML
TCD0277-25MLEA
25.37
USD
InStock
TCD0277-25ML
TCD0277-500ML
1,2-Diaminocyclohexane ≥95.0%
1,2-Diaminocyclohexane
CAS Number: 694-83-7
MDL Number: MFCD00001491
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 376
Flash Point (°C): 75
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: 1,2-Diaminocyclohexane