Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2-Dichloroacetamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

D0306-25G
TCD0306-025GEA 184.73 USD InStock
TCD0306-025G
2,2-Dichloroacetamide ≥98.0% (by total nitrogen basis)
2,2-Dichloroacetamide
CAS Number: 683-72-7
MDL Number: MFCD00008015
Molecular Formula: C2H3Cl2NO
Molecular Weight: 127.95
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Boiling point (°C): 234
Melting point (°C): 99


Formula: C₂H₃Cl₂NO
MW: 127.96 g/mol
Boiling Pt: 234 °C
Melting Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00008015
CAS Number: 683-72-7
EINECS: 211-674-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/68/28974768.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty