Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dichloroacetone ≥98.0%

Supplier: TCI America

D0401-25G D0401-500G
TCD0401-500GEA 377.41 USD InStock
TCD0401-500G TCD0401-25G
1,3-Dichloroacetone ≥98.0%
1,3-Dichloroacetone
CAS Number: 534-07-6
MDL Number: MFCD00000937
Molecular Formula: C3H4Cl2O
Molecular Weight: 126.96
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 173
Melting point (°C): 43
Flash Point (°C): 124

Other Product Names / Synonyms: 1,3-Dichloroacetone


Formula: ClH₂CC(O)CH₂Cl
MW: 126.97 g/mol
Boiling Pt: 173 °C
Melting Pt: 43 °C
Density: 1.432 g/cm³ (20 °C)
Flash Pt: 124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000937
CAS Number: 534-07-6
EINECS: 208-585-6
UN: 2649
ADR: 6.1,II
Merck Index: 13,03077
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/19/28974919.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty