Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dichlorotoluene ≥98.0%

Supplier: TCI America
Synonyms: 1,2-Dichloro-4-methylbenzene

D0426-500G D0426-25G
TCD0426-025GEA 30.45 USD InStock
TCD0426-025G TCD0426-500G
3,4-Dichlorotoluene ≥98.0%
3,4-Dichlorotoluene
CAS Number: 95-75-0
MDL Number: MFCD00000556
Molecular Formula: C7H6Cl2
Molecular Weight: 161.03
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 118
Specific Gravity (20/20): 1.25


Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 205 °C
Melting Pt: –15.2 °C
Density: 1.25 (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00000556
CAS Number: 95-75-0
EINECS: 202-447-9
UN: 3082
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/60/28974960.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty