Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N,N-diethylacetamide ≥98.0%

Supplier: TCI America
Synonyms: 2-Chloro-n,n-diethylacetamideN,N,-Diethylchloroacetamide

D0483-25G D0483-500G
TCD0483-25GEA 43.59 USD InStock
TCD0483-25G TCD0483-500G
2-Chloro-N,N-diethylacetamide ≥98.0%
2-Chloro-N,N-diethylacetamide
CAS Number: 2315-36-8
MDL Number: MFCD00000928
Molecular Formula: C6H12ClNO
Molecular Weight: 149.62
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Specific Gravity (20/20): 1.09

Other Product Names / Synonyms: N-Chloroacetyldiethylamine


Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
Boiling Pt: 150 °C
Density: 1.09 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000928
CAS Number: 2315-36-8
EINECS: 219-019-2
UN: 2810
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/50/58/28975058.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty