Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane ≥99.5% stabilized

Supplier: TCI America
Warning
Synonyms: Methane dichlorideMethylene chloride

Stabilization: Stabilised with Stabilised

D0529-100ML
TCD0529-100MLEA 63.64 USD InStock
TCD0529-100ML
Dichloromethane ≥99.5% stabilized
Dichloromethane
(stabilized with 2-Methyl-2-butene)
[for Spectrophotometry]
CAS Number: 75-09-2
MDL Number: MFCD00000881
Molecular Formula: CH2Cl2
Molecular Weight: 84.93
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 39
Melting point (°C): -97
Specific Gravity (20/20): 1.33

Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Methylene Chloride


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39 °C
Melting Pt: –97 °C
Density: 1.33 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.5% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty