Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dihydroxybenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0568-25G
TCD0568-025GEA 34.51 USD InStock
TCD0568-025G
2,4-Dihydroxybenzoic acid ≥98.0% (by titrimetric analysis)
2,4-Dihydroxybenzoic acid
CAS Number: 89-86-1
MDL Number: MFCD00002451
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 218
Flash Point (°C): 149

Other Product Names / Synonyms: beta-Resorcylic Acid


Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 390 °C (1013 hPa)
Melting Pt: 218 °C
Density: 1.54 g/cm³ (20 °C)
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00002451
CAS Number: 89-86-1
EINECS: 201-946-9
Merck Index: 12,08325
https://us-legacy.vwr.com/stibo/web/std.lang.all/52/38/28975238.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty