Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,5-Dinitronaphthalene ≥98.0%

Supplier: TCI America

D0836-25G D0836-250G
TCD0836-025GEA 46.69 USD InStock
TCD0836-025G TCD0836-250G
1,5-Dinitronaphthalene ≥98.0%
1,5-Dinitronaphthalene
CAS Number: 605-71-0
MDL Number: MFCD00003916
Molecular Formula: C10H6N2O4
Molecular Weight: 218.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 218


Formula: C₁₀H₆N₂O₄
MW: 218.17 g/mol
Boiling Pt: 390 °C (1013 hPa)
Melting Pt: 218 °C
Density: 1.586 g/cm³ (20 °C)
MDL Number: MFCD00003916
CAS Number: 605-71-0
EINECS: 210-095-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/59/34/28975934.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty