Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyltin dichloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger
Synonyms: Dichlorodimethylstannane

D1338-500G D1338-25G D1338-100G
TCD1338-100GEA 196.82 USD InStock
TCD1338-100G TCD1338-500G TCD1338-025G
Dimethyltin dichloride ≥99.0% (by titrimetric analysis)
Dimethyltin dichloride
CAS Number: 753-73-1
MDL Number: MFCD00000517
Molecular Formula: C2H6Cl2Sn
Molecular Weight: 219.68
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 107


Formula: C₂H₆Cl₂Sn
MW: 219.69 g/mol
Boiling Pt: 180 °C
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 3146
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/82/05/28978205.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty