Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Diphenylpropane ≥95.0%

Supplier: TCI America

D1412-5ML D1412-25ML
TCD1412-25MLEA 497.49 USD InStock
TCD1412-25ML TCD1412-005ML
1,3-Diphenylpropane ≥95.0%
1,3-Diphenylpropane
CAS Number: 1081-75-0
MDL Number: MFCD00043574
Molecular Formula: C15H16
Molecular Weight: 196.29
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Specific Gravity (20/20): 0.98


Formula: (C₆H₅)CH₂CH₂CH₂(C₆H₅)
MW: 196.29 g/mol
Boiling Pt: 83 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00043574
CAS Number: 1081-75-0
EINECS: 214-101-4
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty