Synonyms:
(±)-Decan-5-olide
, 5-Decanolide
, 6-Pentyltetrahydro-2H-pyran-2-one
, (±)-Tetrahydro-6-pentyl-2H-pyran-2-one
D1416-25ML
TCD1416-025MLEA
53.9
USD
InStock
TCD1416-025ML
(±)-δ-Decalactone ≥97.0%
(±)-δ-Decalactone
CAS Number: 705-86-2
MDL Number: MFCD00006649
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 145
Specific Gravity (20/20): 0.97
Product Categories: Materials Chemistry, Monomers, Bioscience, Lipids
Other Product Names / Synonyms: Amyl-delta-valerolactone, delta-Caprinolactone