Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Di-tert-butyl-4-hydroxybenzyl alcohol ≥97.0%

Supplier: TCI America

D1480-25G
TCD1480-025GEA 243.97 USD InStock
TCD1480-025G
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol ≥97.0%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol
CAS Number: 88-26-6
MDL Number: MFCD00017254
Molecular Formula: C15H24O2
Molecular Weight: 236.36
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 140
Flash Point (°C): 144


Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Melting Pt: 140 °C
Flash Pt: 144 °C
MDL Number: MFCD00017254
CAS Number: 88-26-6
EINECS: 201-815-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/06/28978306.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty