Synonyms:
1,3-Dibromo-2,4,5,6-tetrafluorobenzene
D1492-5G
D1492-1G
TCD1492-1GEA
49.66
USD
InStock
TCD1492-1G
TCD1492-005G
1,3-Dibromotetrafluorobenzene ≥98.0%
1,3-Dibromotetrafluorobenzene
CAS Number: 1559-87-1
MDL Number: MFCD00031533
Molecular Formula: C6Br2F4
Molecular Weight: 307.87
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 92
Specific Gravity (20/20): 2.24