Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,5-Diphenylcarbazide ≥98.0% (by HPLC)

Supplier: TCI America
Warning
Synonyms: sym. Diphenylcarbazidesym-DiphenylcarbazideN'',N'''-Diphenylcarbonohydrazide1,5-DiphenylcarbohydrazideDiphenylcarbazide1,5-Diphenylcarbonohydrazide

D1513-25G
TCD1513-025GEA 94.03 USD InStock
TCD1513-025G
1,5-Diphenylcarbazide ≥98.0% (by HPLC)
1,5-Diphenylcarbazide
CAS Number: 140-22-7
MDL Number: MFCD00003013
Molecular Formula: C13H14N4O
Molecular Weight: 242.28
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 175
Storage Temperature: 0°C

Other Product Names / Synonyms: 1,5-Diphenylcarbazide


Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 454 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/28/28978328.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty