Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dibromopyridine ≥98.0%

Supplier: TCI America

D1555-250G D1555-25G
TCD1555-025GEA 44.84 USD InStock
TCD1555-025G TCD1555-250G
2,6-Dibromopyridine ≥98.0%
2,6-Dibromopyridine
CAS Number: 626-05-1
MDL Number: MFCD00006223
Molecular Formula: C5H3Br2N
Molecular Weight: 236.89
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 249
Melting point (°C): 119
Flash Point (°C): 100


Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 249 °C
Melting Pt: 119 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006223
CAS Number: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/62/28978362.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty