Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decafluorobenzophenone ≥98.0%

Supplier: TCI America
Synonyms: Perfluorobenzophenone

D1631-1G D1631-5G
TCD1631-001GEA 81.12 USD InStock
TCD1631-001G TCD1631-005G
Decafluorobenzophenone ≥98.0%
Decafluorobenzophenone
CAS Number: 853-39-4
MDL Number: MFCD00000295
Molecular Formula: C13F10O
Molecular Weight: 362.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 93

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluorobenzophenone


Formula: C₁₃F₁₀O
MW: 362.13 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000295
CAS Number: 853-39-4
EINECS: 212-717-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/84/25/28978425.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty