Synonyms:
(2,4-Dichlorophenyl)methanol
D1837-25G
TCD1837-025GEA
52
USD
InStock
TCD1837-025G
2,4-Dichlorobenzyl alcohol ≥97.0%
2,4-Dichlorobenzyl alcohol
CAS Number: 1777-82-8
MDL Number: MFCD00004606
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 59