Synonyms:
Pentamethylene diiodide
Stabilization:
Stabilised with Stabilised
D2110-25G
TCD2110-025GEA
133.98
USD
InStock
TCD2110-025G
1,5-Diiodopentane ≥98.0% stabilized
1,5-Diiodopentane
(stabilized with Copper chip)
CAS Number: 628-77-3
MDL Number: MFCD00001101
Molecular Formula: C5H10I2
Molecular Weight: 323.94
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 110
Freezing point (°C): 9
Specific Gravity (20/20): 2.18
Other Product Names / Synonyms: Pentamethylene Diiodide