Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloroacetonitrile ≥98.0%

Supplier: TCI America

D2510-25G D2510-500G
TCD2510-25GEA 73 USD InStock
TCD2510-25G TCD2510-500G
Dichloroacetonitrile ≥98.0%
Dichloroacetonitrile
CAS Number: 3018-12-0
MDL Number: MFCD00040886
Molecular Formula: C2HCl2N
Molecular Weight: 109.94
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 35
Specific Gravity (20/20): 1.37


Formula: C₂HCl₂N
MW: 109.94 g/mol
Boiling Pt: 112 °C
Density: 1.37 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00040886
CAS Number: 3018-12-0
EINECS: 221-159-4
UN: 2920
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/94/46/28979446.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty